- Determination of molybdenum in environmental samples.
Determination of molybdenum in environmental samples.
Analytica chimica acta (2007-04-10)
Krystyna Pyrzynska
PMID17416221
摘要
Determination of molybdenum in different kinds of environmental samples is often a challenging task for analysts. Its concentration is usually very low and the sample matrix may cause serious interferences during measurement. Therefore, preconcentration and separation methods should be used to solve these problems and render more sensitive, accurate and interference-free determination. Recent developments in sample treatment, such as solid phase and liquid-liquid extraction as well as coprecipitation are presented, including flow-based methodology. In addition, important extension and improvements in analytical methods for determinations of molybdenum are updated. Some examples of speciation analysis are also presented.
材料
货号
品牌
产品描述
钼, foil, 15mm disks, thickness 0.005mm, 99.9%
钼, foil, 25mm disks, thickness 0.0125mm, 99.9%
钼, rod, 200mm, diameter 6.0mm, centerless ground, 99.9%
钼, pellets, 100g, max. size 10mm, 99.9%
钼, foil, 150x150mm, thickness 1.0mm, annealed, 99.98%
钼, wire, straight, 1000mm, diameter 0.25mm, 99.95%
钼, foil, light tested, 100x100mm, thickness 0.01mm, 99.9%
钼, pellets, 200g, max. size 10mm, 99.9%
钼, rod, 50mm, diameter 5.0mm, 99.98%
钼, mesh, 100x100mm, nominal aperture 0.44mm, wire diameter 0.07mm, 50 wires/inch, open area 67%, plain weave mesh
钼, pellets, 50g, max. size 10mm, 99.9%
钼, tube, 100mm, outside diameter 1.0mm, inside diameter 0.7mm, wall thickness 0.15mm, 99.9%
钼, rod, 100mm, diameter 2.0mm, centerless ground, 99.9%
钼, tube, 50mm, outside diameter 6.4mm, inside diameter 4.6mm, wall thickness 0.9mm, 99.9%
钼, foil, light tested, 50x50mm, thickness 0.01mm, 99.9%
钼, rod, 200mm, diameter 2.0mm, centerless ground, 99.9%
钼, foil, 100x100mm, thickness 0.5mm, annealed, 99.9%
钼, foil, 1m coil, thickness 0.01mm, 99.9%
钼, foil, light tested, 130x130mm, thickness 0.015mm, as rolled, 99.9%
钼, insulated wire, 10m, conductor diameter 0.125mm, insulation thickness 0.012mm, polyimide insulation, 99.95%
钼, foil, 100x100mm, thickness 0.30mm, annealed, 99.9%
钼, wire, straight, 1000mm, diameter 0.5mm, 99.95%
钼, foil, 25x25mm, thickness 0.125mm, annealed, 99.9%
钼, rod, 150mm, diameter 5.0mm, 99.98%
钼, rod, 200mm, diameter 5.0mm, centerless ground, 99.9%