跳转至内容
Merck
  • [Implication of 5-HT2A receptors in the genetic mechanisms of the brain 5-HT system autoregulation].
[Implication of 5-HT2A receptors in the genetic mechanisms of the brain 5-HT system autoregulation].
Molekuliarnaia biologiia (2012-08-15)
V S Naumenko, A S Tsybko, D V Bazovkina, N K Popova
摘要
Brain serotonin (5-HT) system has been implicated in pathophysiology of anxiety, depression, drug addiction, and schizophrenia. 5-HT2A receptor is involved in the mechanisms of stress-induced psychopathology and impulsive behavior. Here, we investigated the role of 5-HT2A receptor in the autoregulation of the brain 5-HT system. The chronic treatment with agonist of 5-HT2A receptor DOI (1.0 mg/kg, i.p./14 days) produced considerable decrease of 5-HT2A receptor-mediated "head-twitches" in AKR/J mice indicating desensitization of 5-HT2A receptors. Chronic DOI treatment failed to alter 5-HT2A receptor gene expression in the midbrain, hippocampus and frontal cortex. At the same time, the increase in the expression of the gene encoding key enzyme of 5-HT synthesis, tryptophan hydroxylase 2 (TPH2), the increase in TPH2 activity and 5-HT levels and decreased expression of serotonin transporter (5-HTT) gene was found in the midbrain of DOI-treated mice. The results provide new evidence of receptor-gene cross-talk in the brain 5-HT system and the implication of 5-HT2A receptor in the autoregulation of the brain 5-HT system.
材料
货号
品牌
产品描述
Sigma-Aldrich
(R)(−)-DOI hydrochloride, ≥98% (HPLC), solid