跳转至内容
Merck
  • C2-symmetric bisprolinamide as a highly efficient catalyst for direct aldol reaction.
C2-symmetric bisprolinamide as a highly efficient catalyst for direct aldol reaction.
Organic letters (2005-11-05)
Sampak Samanta, Jinyun Liu, Rajasekhar Dodda, Cong-Gui Zhao
摘要
[reaction: see text] The catalytic activity of the prolinamide-type catalysts may be improved by introducing additional prolinamide moiety into the catalyst, while the enantioselectivity can still be maintained or further improved. A C2-symmetric bisprolinamide with two prolinamide moieties has been found to be an excellent catalyst for direct aldol reaction with more than doubled reactivity and better asymmetric induction than its monoprolinamide counterpart.
材料
货号
品牌
产品描述
Sigma-Aldrich
L-脯氨酰胺, 98%