跳转至内容
Merck
  • Temperature-programmable low thermal mass silicon micromachined gas chromatography and differential mobility detection for the fast analysis of trace level of ethylene oxide in medical work place atmospheres.
Temperature-programmable low thermal mass silicon micromachined gas chromatography and differential mobility detection for the fast analysis of trace level of ethylene oxide in medical work place atmospheres.
Journal of chromatography. A (2012-07-20)
J Luong, R Gras, H J Cortes, R A Shellie
摘要
A portable, fast gas chromatographic method for the direct measurement of part-per-billion level of ethylene oxide in medical work place atmospheres is introduced. Ethylene oxide is a chemical of significance in life science for its critical role as a highly effective sterilizing agent for heat sensitive surgical instruments. However, ethylene oxide is highly flammable, a suspected human carcinogen, is mutagenic, and henceforth, requires close monitoring. The approach incorporates a temperature-programmable silicon micromachined gas chromatograph for the fast separation of ethylene oxide from airborne organic interferences. The separation approach is hyphenated to a micromachined differential mobility detector, improving targeted analyte detection, and enhancing the overall selectivity obtained. A complete analysis can be conducted in less than 60s. Further, ethylene oxide in the matrix mentioned can be measured directly with low possibility of false positives and without the need for any sample pre-treatment, such as pre-concentration or derivatization. A relative precision of less than 5% RSD (n=20) over a range from 5 parts per billion (v/v) to 500 parts per billion (v/v) was obtained.
材料
货号
品牌
产品描述
Supelco
环氧乙烷 溶液, certified reference material, 50 mg/mL in methanol
Sigma-Aldrich
环氧乙烷 溶液, 2.5-3.3 M in THF