跳转至内容
Merck
  • [Determination of methanol and fusel oils in alcohol beverages using headspace solid-phase microextraction and gas chromatography].
[Determination of methanol and fusel oils in alcohol beverages using headspace solid-phase microextraction and gas chromatography].
Se pu = Chinese journal of chromatography (2003-01-25)
Hong-he Liu, Yuan-qian Li, Cheng-jun Sun
摘要
A method for the determination of methanol and fusel oils in alcohol beverages using headspace solid-phase microextraction and gas chromatography (HS-SPME-GC) is presented. The solid phase was a coated epoxy resin. The extraction and chromatography conditions were optimized. Limits of detection were 0.02 mg/L-0.04 mg/L and relative standard deviations were in the range of 1.4%-4.1%. The proposed method showed better sensitivity in comparing with direct headspace gas chromatography(HS-GC, the National Standard method). This method was applied to evaluate real samples. The spiked recoveries in beer, wine and functional alcohol samples ranged from 80.8% to 110.3% for methanol and fusel oils. The results by HS-SPME-GC and HS-GC for alcohol samples coincided very well. The proposed method is simple, fast and accurate with high reproducibility, high sensitivity and low cost. It extends the applications of SPME.
材料
货号
品牌
产品描述
Sigma-Aldrich
Fusel oil, natural, FG