跳转至内容
Merck
Fate of isoxaflutole in soil under controlled conditions.
Journal of agricultural and food chemistry (2002-12-28)
Estelle Beltrán, Hélène Fenet, Jean-Françoise Cooper, Camille-Michel Coste
摘要
Isoxaflutole (IFT, 5-cyclopropyl-1,2-oxazol-4yl-alpha,alpha,alpha-trifluoro-2-mesyl-p-tolyl ketone) is a new pre-emergence proherbicide used in maize and sugarcane. Its two main derivatives are a diketonitrile derivative, 2-cyano-3-cyclopropyl-1-(2-methanesulfonyl-4-trifluoromethylphenyl)propane-1,3-dione, called DKN, and a benzoic acid derivative, 2-methanesulfonyl-4-trifluoromethylbenzoic acid, called BA. Few data are available of the factors influencing the degradation of IFT in soil, and the purpose of the present work was to determine the relative importance of, and factors affecting, the degradation of IFT in soil. Experiments were conducted on five soils with distinct physicochemical characteristics, at different temperatures and moisture contents in biotic and abiotic conditions. The isomerization of IFT to DKN is rapid, increasing with higher moisture contents and higher temperatures. It depends strongly on pH and is governed by chemical processes. The degradation of DKN to BA appeared to be essentially due to the biological activity of the soil.
材料
货号
品牌
产品描述
Supelco
异恶唑草酮, PESTANAL®, analytical standard