跳转至内容
Merck
  • Initiation and disassembly of filopodia tip complexes containing VASP and lamellipodin.
Initiation and disassembly of filopodia tip complexes containing VASP and lamellipodin.
Molecular biology of the cell (2020-06-25)
Karen W Cheng, R Dyche Mullins
摘要
The shapes of many eukaryotic cells depends on the actin cytoskeleton, and changes in actin assembly dynamics underlie many changes in cell shape. Ena/VASP-family actin polymerases, for example, modulate cell shape by accelerating actin filament assembly locally and slowing filament capping. When concentrated into discrete foci at the leading edge, VASP promotes filopodia assembly and forms part of a poorly understood molecular complex that remains associated with growing filopodia tips. Here we identify precursors of this filopodia tip complex in migrating B16F1 cells: small leading-edge clusters of the adaptor protein lamellipodin (Lpd) that subsequently recruit VASP and initiate filopodia formation. Dimerization, membrane association, and VASP binding are all required for lamellipodin to incorporate into filopodia tip complexes, and overexpression of monomeric, membrane--targeted lamellipodin mutants disrupts tip complex assembly. Once formed, tip complexes containing VASP and lamellipodin grow by fusing with each other, but their growth is limited by a size-dependent dynamic instability. Our results demonstrate that assembly and disassembly dynamics of filopodia tip complexes are determined, in part, by a network of multivalent interactions between Ena/VASP proteins, EVH1 ligands, and actin filaments.
材料
货号
品牌
产品描述
Sigma-Aldrich
层粘连蛋白 来源于 Engelbreth-Holm-Swarm 小鼠肉瘤基底膜, 1-2 mg/mL in Tris-buffered saline, 0.2 μm filtered, BioReagent, suitable for cell culture
货号
包装规格
是否有货
价格
数量
Sigma-Aldrich
细胞松驰素D, from Zygosporium mansonii, ≥98% (TLC and HPLC), powder
货号
包装规格
是否有货
价格
数量