跳转至内容
Merck
  • TRIM28 functions as the SUMO E3 ligase for PCNA in prevention of transcription induced DNA breaks.
TRIM28 functions as the SUMO E3 ligase for PCNA in prevention of transcription induced DNA breaks.
Proceedings of the National Academy of Sciences of the United States of America (2020-09-10)
Min Li, Xiaohua Xu, Chou-Wei Chang, Yilun Liu
摘要
In human cells, the DNA replication factor proliferating cell nuclear antigen (PCNA) can be conjugated to either the small ubiquitinlike modifier SUMO1 or SUMO2, but only SUMO2-conjugated PCNA is induced by transcription to facilitate resolution of transcription-replication conflict (TRC). To date, the SUMO E3 ligase that provides substrate specificity for SUMO2-PCNA conjugation in response to TRC remains unknown. Using a proteomic approach, we identified TRIM28 as the E3 ligase that catalyzes SUMO2-PCNA conjugation. In vitro, TRIM28, together with the RNA polymerase II (RNAPII)-interacting protein RECQ5, promotes SUMO2-PCNA conjugation but inhibits SUMO1-PCNA formation. This activity requires a PCNA-interacting protein (PIP) motif located within the bromodomain of TRIM28. In cells, TRIM28 interaction with PCNA on human chromatin is dependent on both transcription and RECQ5, and SUMO2-PCNA level correlates with TRIM28 expression. As a consequence, TRIM28 depletion led to RNAPII accumulation at TRC sites, and expression of a TRIM28 PIP mutant failed to suppress TRC-induced DNA breaks.
材料
货号
品牌
产品描述
Sigma-Aldrich
MISSION® esiRNA, targeting human TRIM28