跳转至内容
Merck
Molecular evolution and structure of alpha-actinin.
Molecular biology and evolution (2004-03-12)
Ana Virel, Lars Backman
摘要
The N-terminal actin-binding domain of alpha-actinin is connected to the C-terminal EF-hands by a rod domain. Because of its ability to form dimers, alpha-actinin can cross-link actin filaments in muscle cells as well as in nonmuscle cells. In the prototypic alpha-actinins, the rod domain contains four triple helical bundles, or so-called spectrin repeats. We have found some atypical alpha-actinins in early diverging organisms, such as protozoa and yeast, where the rod domain contains one and two spectrin repeats, respectively. This implies that the four repeats present in modern alpha-actinins arose after two consecutive intragenic duplications from an alpha-actinin with a single repeat. Further, the evolutionary gene tree of alpha-actinins shows that the appearance of four distinct alpha-actinin isoforms may have occurred after the vertebrate-invertebrate split. The topology of the tree lends support to the hypothesis that two rounds (2R) of genome duplication occurred early in the vertebrate radiation. The phylogeny also considers these atypical isoforms as the most basal to alpha-actinins of vertebrates and other eukaryotes. The analysis also positioned alpha-actinin of the fungi Encephalitozoo cuniculi close to the protozoa, supporting the suggestion that microsporidia are early eukaryotes. Because alpha-actinin is considered the basal member of the spectrin family, our studies will improve the understanding of the origin and evolution of this superfamily.