跳转至内容
Merck
  • Human DECR1 is an androgen-repressed survival factor that regulates PUFA oxidation to protect prostate tumor cells from ferroptosis.
Human DECR1 is an androgen-repressed survival factor that regulates PUFA oxidation to protect prostate tumor cells from ferroptosis.
eLife (2020-07-21)
Zeyad D Nassar, Chui Yan Mah, Jonas Dehairs, Ingrid Jg Burvenich, Swati Irani, Margaret M Centenera, Madison Helm, Raj K Shrestha, Max Moldovan, Anthony S Don, Jeff Holst, Andrew M Scott, Lisa G Horvath, David J Lynn, Luke A Selth, Andrew J Hoy, Johannes V Swinnen, Lisa M Butler
摘要
Fatty acid β-oxidation (FAO) is the main bioenergetic pathway in human prostate cancer (PCa) and a promising novel therapeutic vulnerability. Here we demonstrate therapeutic efficacy of targeting FAO in clinical prostate tumors cultured ex vivo, and identify DECR1, encoding the rate-limiting enzyme for oxidation of polyunsaturated fatty acids (PUFAs), as robustly overexpressed in PCa tissues and associated with shorter relapse-free survival. DECR1 is a negatively-regulated androgen receptor (AR) target gene and, therefore, may promote PCa cell survival and resistance to AR targeting therapeutics. DECR1 knockdown selectively inhibited β-oxidation of PUFAs, inhibited proliferation and migration of PCa cells, including treatment resistant lines, and suppressed tumor cell proliferation and metastasis in mouse xenograft models. Mechanistically, targeting of DECR1 caused cellular accumulation of PUFAs, enhanced mitochondrial oxidative stress and lipid peroxidation, and induced ferroptosis. These findings implicate PUFA oxidation via DECR1 as an unexplored facet of FAO that promotes survival of PCa cells.
材料
货号
品牌
产品描述
Sigma-Aldrich
单克隆抗 β-肌动蛋白抗体 小鼠抗, clone AC-15, ascites fluid
Sigma-Aldrich
亚油酸, ≥98%
Sigma-Aldrich
铁抑制剂-1, ≥95% (HPLC)
Sigma-Aldrich
(+)-依托莫西 钠盐 水合物, ≥98% (HPLC), powder
Sigma-Aldrich
Anti-DECR1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab3