跳转至内容
Merck
  • Dissipation Dynamics and Residue of Four Herbicides in Paddy Fields Using HPLC-MS/MS and GC-MS.
Dissipation Dynamics and Residue of Four Herbicides in Paddy Fields Using HPLC-MS/MS and GC-MS.
International journal of environmental research and public health (2019-01-18)
Qian Yu, Ping Zhang, Yuhan He, Zhifeng Xu, Xiulong He, Yuan Hu, Hongjun Zhang, Lin He
摘要
The dissipation dynamics and residue of pyrazosulfuron-ethyl, bensulfuron-methyl, acetochlor, and butachlor in paddy fields at Good Agricultural Practices (GAP) condition were carefully investigated in this study. The four herbicides' residues were determined based on a quick, easy, cheap, rugged, safe (QuEChERS) method coupled with HPLC-MS/MS and GC-MS. The limit of detection (LOD) for pyrazosulfuron-ethyl, bensulfuron-methyl, acetochlor, and butachlor in all matrices ranged from 0.04⁻1.0 ng. The limit of quantification (LOQ) of the four herbicides ranged from 0.01⁻0.1 mg/kg. Moreover, the average recoveries of the four herbicides ranged from 78.9⁻108% with relative standard deviations (RSDs) less than 15% at three different fortified levels for different matrices. The dissipation results indicated that the average half-lives (t1/2) of the four herbicides in soil were in the range of 3.5⁻17.8 days, and more than 95% of the four herbicides dissipated within 5 days in water. Furthermore, the final residues of the four herbicides were all below the LOQ at harvest time. Such results highlight the dissipation dynamics and residue of the four herbicides in a rice cropping system and contribute to risk assessment as well as scientific guidance on the proper and safe application of herbicides in paddy fields.