返回
PTFE washer for Quickfit
®
cone/screw thread adapter
Products
产品分类
washer (6)
制造商
筛选产品
Showing 1-6 of 6
比较
货号
说明
价格
Z320196
PTFE washer for Quickfit
®
cone/screw thread adapter,
13 thread, couples 6.0 - 7.0 mm diameter
展开
查看定价
Z320218
PTFE washer for Quickfit
®
cone/screw thread adapter,
18 thread, couples 7.0 - 8.5 mm diameter
展开
查看定价
Z320226
PTFE washer for Quickfit
®
cone/screw thread adapter,
24 thread, couples 6.0 - 7.0 mm diameter
展开
查看定价
Z320242
PTFE washer for Quickfit
®
cone/screw thread adapter,
24 thread, couples 10.5 - 11.5 mm diameter
展开
查看定价
Z320269
PTFE washer for Quickfit
®
cone/screw thread adapter,
28 thread, couples 10.5 - 11.5 mm diameter
展开
查看定价
Z320277
PTFE washer for Quickfit
®
cone/screw thread adapter,
28 thread, couples 18.0 - 19.0 mm diameter