返回
玻璃液体闪烁瓶
Products
产品分类
vials (5)
品牌
Supelco (2)
容量 (ml)
闭合类型
小瓶装材料
筛选产品
Showing 1-5 of 5
比较
货号
说明
价格
V7130
玻璃液体闪烁瓶,
volume 20 mL, vial size 28 mm × 61 mm , with cap, the cap is made of urea and the liner material is a polyethylene cone, pkg of 500 ea
展开
查看定价
V8255
玻璃液体闪烁瓶,
volume 20 mL, vial size 28 mm × 61 mm , with cap, Cap: 22mm urea with metal foil liner. (caps packed separately), pkg of 500 ea
展开
查看定价
V7005
玻璃液体闪烁瓶,
volume 20 mL, vial size 28 mm × 61 mm , with cap, Cap: 22mm urea with Poly-Seal cone liner. (caps packed separately), pkg of 500 ea
展开
查看定价
M1901
玻璃液体闪烁瓶,
volume 6 mL, Minivial, vial size 17 mm × 58 mm , with cap, Cap: 15mm urea with metal foil liner., pkg of 1000 ea
展开
查看定价
M1152
玻璃液体闪烁瓶,
volume 6 mL, Minivial, vial size 17 mm × 58 mm , with cap, Cap: 13mm urea with metal foil liner., pkg of 1000 ea