返回
Hellma
®
cuvettes, demountable cells
Products
产品分类
cuvettes (2)
制造商
体积(µL)
检测方法
筛选产品
Showing 1-2 of 2
比较
货号
说明
价格
Z800058
Hellma
®
cuvettes, demountable cells,
High Performance Quartz Glass, spectral range 200-2500 nm, pathlength 0.5 ± 0.010 mm, chamber volume 130 μL
展开
查看定价
Z805963
Hellma
®
cuvettes, demountable cells,
High Performance Quartz Glass, spectral range 200-2500 nm, pathlength 0.1 ± 0.005 mm, chamber volume 26 μL