返回
Ace replacement stirrer shaft with agitator, PTFE coated SS
Products
制造商
直径(mm)
包装大小
筛选产品
Showing 1-1 of 1
比较
货号
说明
价格
Z690589
Ace replacement stirrer shaft with agitator, PTFE coated SS,
diam. 10 mm, fits 1 L PTFE reactors