主要文件
K5164-U
Omnifit® EZ Benchmark Replacement Glass Columns
I.D. × L 15 mm × 400 mm
登录查看公司和协议定价
选择尺寸
1 EA
$255.00
$255.00
预计发货时间2025年6月06日详情
获取大包装报价
选择尺寸
变更视图
1 EA
$255.00
About This Item
分類程式碼代碼:
23151816
NACRES:
SB.52
$255.00
预计发货时间2025年6月06日详情
获取大包装报价
製造商/商標名
Omnifit 006ERG-15-40
品質等級
技術
LPLC: suitable
內徑 × 長度
15 mm × 400 mm
正在寻找类似产品? 访问 产品对比指南
一般說明
Replacement glass columns are available in a wide range of bore sizes and lengths. This not only enables you to replace a column should breakage occur, but allows you to obtain several different length columns to use with endpieces you already have for that bore size. The modular nature of our column ranges means that for the ultimate flexibility you can mix and match column sizes with different endpiece types to your specific requirements.
法律資訊
Omnifit is a registered trademark of Diba Industries
历史批次信息供参考:
分析证书(COA)