Key Documents
EP5490041003
Rotor for Eppendorf® Concentrator Plus
F-50-8-16, non aerosol tight, to hold, 8 x 8.0/12.0 mL round-bottom tubes
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41103909
NACRES:
NB.82
特點
to hold 8 x 16 x 105 -120 mm
to hold 8 x 8.0/12.0 mL round-bottom tubes
fixed angle rotor
non aerosol tight
包裝
pack of 1 ea
製造商/商標名
Eppendorf® 5490041003 (EU)
技術
centrifugation: suitable
相容性
for use with centrifuge models Concentrator Plus
正在寻找类似产品? 访问 产品对比指南
法律資訊
Eppendorf is a registered trademark of Eppendorf AG