Merck
HomeCancer ResearchAtto Dyes for Superior Fluorescent Imaging
Atto Dyes for Superior Fluorescent Imaging
Activated fluorescent dyes are routinely used to tag proteins, nucleic acids, and other biomolecules for use in life science applications including fluorescence microscopy, flow cytometry, fluorescence in situ hybridization (FISH), receptor binding assays, and enzyme assays. The Atto dyes are a series of fluorescent dyes that meet the critical needs of modern fluorescent technologies:
  • Stability - Atto 655 and Atto 647N are photostable and highly resistant to ozone degradation, making them ideal for microarray applications. See the related article "Analyzing Properties of Fluorescent Dyes used for Labeling DNA in Microarray Experiments".
  • Long Signal Lifetimes - Signal decay times of 0.6–4.1 nanoseconds allow timegate studies to reduce autofluorescence background and scattering.
  • Reduced Background - Several Atto dyes have excitation wavelengths greater than 600 nm, reducing background fluorescence from samples, Rayleigh and Raman scattering.
  • Selection - Atto dyes have strong fluorescent signals that cover visible and near-IR emission wavelengths.
Atto Dye's Long Signal Lifetimes
Atto dyes exhibit longer fluorescence signal lifetimes (0.6–4.1 ns) in aqueous solution than either carbocyanine dyes or most of the autofluorescence inherent in cells and biomolecules. The signal from Atto dyes can be measured using pulsed laser excitation with a time-gated detection system to reduce interference from fluorophores with shorter lifetimes, background autofluorescence, and Rayleigh and Raman light scattering, improving overall sensitivity.
Long Excitation Wavelengths Reduces Background
Diode laser excitation at 635 nm and redabsorbing fluorescent dyes were shown to reduce autofluorescence of biological samples sufficiently so that individual antigen and antibody molecules could be detected in human serum samples.1,2 Excitation in the red spectral region also reduces cell damage when working with live cells.3
Many of Atto dyes (Atto 590 and above) can be excited using wavelengths greater than 600 nm. Using long-wavelength activated Atto dyes in conjunction with the appropriate excitation wavelength reduces autofluorescence due to sample, solvent, glass, or polymer support, and improves overall sensitivity in biological analysis and imaging techniques. The background fluorescence due to Rayleigh and Raman scattering are also dramatically reduced by use of longer wavelength excitation.
Emission Wavelength Range from 479 to 764 nm for Fluorescent Multiplex Detection
Atto dyes have strong fluorescent signals with most having molar absorptivity values >100,000 and low excitation/emission overlap, making Atto dyes ideal for multiplex techniques using visible and near-IR emission wavelengths.
With excitation signal maxima ranging from 390 to 740 nm and good Stokes shift separation, there are Atto dyes suitable for use with any common excitation light source.
Fluorescent Multiplex Detection using Atto Dyes
Atto dyes can be used to conjugate probes and biomolecules for multiplex applications. Selection of two Atto dyes with separated emission signals supports multiple excitation and measurement results from a single experiment.
Fluorescent Multiplex Detection using Antibody Atto Dye Conjugates
Immunoblot detection of Protein 1 and Protein 2 using two primary antibodies
Immunoblot detection of Protein 1 and Protein 2 using two primary antibodies and two anti-IgG-Atto dye conjugates. Imaging was done sequentially using a FLA-3000 Fuji® laser scanner, first at an excitation wavelength of 532 nm with a 580 nm emission filter, then at an excitation wavelength of 633 nm with a 675 nm emission filter. The image overlay was done using a software tool.
Alternatives to Common Fluorophores
With the extensive selection of Atto dyes available, any common excitation light source can be used, and Atto dyes can replace other fluorescent dyes commonly used in life science.
FluorophoneRecommended Atto Dye Alternative
Alexa Fluor 488Atto 488
FITCAtto 488
FAM™Atto 488
JOE™Atto 520
TET™Atto 520
Alexa Fluor 532Atto 532
HEX™Atto 532Atto Rho6G
TAMRA™Atto 550
Cy3Atto 550
Cy3.5Atto 565
ROX™Atto 565, Atto Rho11
Alexa Fluor 594Atto 590Atto 594
Texas Red®Atto 590
Alexa Fluor 633Atto 633Atto Rho14
Cy5Atto 647Atto 647NAtto 655
Alexa Fluor 647Atto 647Atto 647NAtto 655
Cy5.5Atto 680Atto 700
Light sourceMain lines (nm)Recommended Atto dyes
Mercury arc lamp365, 405, 436, 546Atto 390, Atto 425, Atto 465, Atto 550, Atto 565
Mercury arc lamp577Atto 590, Atto Rho101, Atto 594; Atto Rho13, Atto 610, Atto 611x
Xenon arc lampContinuum and peaks >800 nmAtto 610, Atto 620, Atto 647, Atto 647N, Atto 655, Atto 680
Halogen lampLittle UV and violet emission; Higher intensity toward longer wavelengthsAtto 610, Atto 620, Atto 647, Atto 647N, Atto 655, Atto 680
Argon ion laser488, 514Atto 488, Atto 520, Atto 532, Atto 550
Argon-krypton laser488, 514, 647, 676Atto 520, Atto 647, Atto 647N, Atto 655, Atto 680
Krypton laser647,676Atto 647, Atto 647N , Atto 655, Atto Oxa12, Atto 665, Atto 680, Atto 700, Atto 725, Atto 740
He-Ne laser633Atto Rho14, Atto 633, Atto 647, Atto 647N
Nd-NAG laser532Atto 532, Atto Rho6G, Atto 550, Atto 565, Atto Rho11, Atto Rho 12
Common diode laser635, 650, 670Atto 633, Atto 647, Atto 647N, Atto 655, Atto 680
Reactive Atto Dyes and Conjugates
Atto dyes produce intense fluorescent signals due to strong absorbance and high quantum yields. Dyes are available in the following formats:
  • Free acid dyes for all routine staining applications
  • NHS-esters for use in common conjugation protocols
  • Maleimides for use in coupling to thiol-containing groups such as cysteine residues and thiol (−SH) tags added during automated synthesis
  • Conjugated to biotin, streptavidin, and antibodies
Atto 655, Atto 680, and Atto 700 are quenched by guanosine, tryptophan and related compounds through direct contact between the dye and the quenching agent and using an electron transfer process. Fluorescent quenching of dyes by tryptophan residues in proteins has been used to differentiate unbound (nonfluorescent) protein from protein-antibody (fluorescent) interactions.1
 λabsε maxλemηemτemCatalog Number
Dyenmm-1  cm-1nm%nsFree AcidNHS EsterMaleimideAzideAmineIodo- acetamideBiotin
Atto 39039024,000479903.889313892048974068321   
Atto 42543645,000484903.556749168054934978948  28616
Atto 430LS43332,000547654.0067145638741882    
Atto 46545375,000508552.2507125340455607    
Atto 48850190,000523803.241051416982856272709744177440230574
Atto 490LS49640,000661302.606715783627836393688   
Atto 49549580,000527452.4169510037941022    
Atto 514511115,000533853.052595674558106244311937288002700713
Atto 520516110,000538903.870706778101659061911  01632
Atto 532532115,000553903.8066998879368499    
Atto Rho6G535115,000560904.1888219351687785   94169
Atto 540Q542105,000   405926168362453    
Atto 550554120,000576803.24242492835307304198639271 28923
Atto 565563120,000592903.475784724641850743069  92637
Atto Rho3B565120,000592501.5936852874376531   07876
Atto Rho11571120,000595804.0566118910151431   19096
Atto Rho12576120,000601804.0444326876172763   91254
Atto Thio12579110,000609152.056963 94079   87784
Atto Rho101586120,000610804.2770855049273522   94379
Atto 580Q586110,000   037224475668152    
Atto 590594120,000624803.7704257963639887   43208
Atto 594601120,000627853.50863708741087177299861583 03927
Atto Rho13600120,000625803.9169736853844535   76141
Atto 610615150,000634703.3784939325941061   43292
Atto 612Q615115,000   043275398850332    
Atto 620619120,000643502.9927166735149728   72978
Atto Rho14625140,000646803.7597466190914397   76140
Atto 633629130,000657643.21862001464n/a685177324568652 
Atto 647645120,000669202.3978750737641784    
Atto 647N644150,000669653.404507183730531691000953497335393606
Atto 655663125,000684301.993711762458066111774149187394806966
Atto Oxa12663125,000684301.8926065578501971   61938
Atto 665663160,000684602.9168510402201407   01376
Atto 680680125,000700301.8948757599904971   55819
Atto 700700120,000719251.53067416986506115982519571 73911
Atto 725729120,000752100.5471569372590979   69616
Atto 740740120,000764100.6913945980877071   50842
Atto MB2658100,000   751187349951797   02934
Convenient Atto Dye Conjugates
An extensive selection of Atto Dye conjugates and kits are available, including:
  • Protein Labeling Kits
    • Atto 488 is a superior alternative to fluorescein and Alexa Fluor 488, producing conjugates with more photostability and brighter fluorescence.
    • Atto 550 is an alternative to rhodamine dyes, Cy3, and Alexa Fluor 550, offering more intense brightness and increased photostability.
    • Atto 594 is an alternative to Alexa Fluor 594 and Texas Red.
    • Atto 647N is an extraordinary highly fluorescent dye, and Atto 655 are alternatives to Cy5 and Alexa Fluor 647.
    • Atto 633 is an alternative to Alexa Fluor 633.
  • Lectins for carbohydrate binding studies.
  • Primary and Secondary Antibodies for direct and indirect ELISA, Immunoblotting, Immunohistochemistry, and other protein identification applications.
  • Biotin and Streptavidin for avidin / streptavidin / biotin conjugation in applications including ELISA, immunohistochemistry, in situ hybridization, and flow cytometry.
Fluorescent microscopy of human skin tissue section
Fluorescent microscopy of human skin tissue section (paraffin fixation) with fungal infection. The target carbohydrate chitotriose of the pathogenic fungi are specifically bound to lectin from Phytolacca americana Atto 488 conjugate (green). The nuclei are counterstained with DAPI (blue). Image by J. Zbären, Inselspital, Bern.
His-tagged p38 MAPK protein
His-tagged p38 MAPK protein (500 ng – 25 ng) was separated on a 4-20% Tris-glycine SDS-PAGE gel. After fixing and washing, the gel was incubated with Ni-NTA-Atto 647N (1:1000) in the dark. The gel was washed and then imaged using a FLA-3000 Fuji® laser scanner with 633 nm excitation and a 675 nm emission filter for Ni-NTA-Atto 647N (λex 647 nm, λem 669 nm). The 50 ng band of His-tagged p38-MAPK is observed using fluorescence imaging.
Atto Dye-labeled Phospholipids
Phospholipids are the major building blocks of biological membranes. The investigation of biological membranes (e.g., intracellular membranes of live cells, plasma membranes) has become a major area of interest. We offer a series of fluorescent-labeled phospholipids. The optical properties of the selected series of dyes allow application with all commonly used excitation and emission filter settings. We offer a variety of phospholipids based on glycerol carrying one or two fatty acids (lipophilic groups) and a phosphate monoester residue (hydrophilic group) such as 1,2-dipalmitoyl-sn-glycero-3-phosphoethanolamine (DPPE), 1,2-dioleoylsn-glycero-3-phosphoethanolamine (DOPE), 1-palmitoyl-2-hydroxy-snglycero-3-phosphoethanolamine (PPE), and 1,2-dimyristoyl-sn-glycero- 3-phosphoethanolamine (DMPE). The fluorophores are covalently linked at the hydrophilic head group of the phospholipids.
Product No.Description
44039Atto 488 DPPE
93580Atto 532 DPPE
51016Atto 550 DPPE
40924Atto 594 DPPE
52816Atto 633 DPPE
05152Atto 647N DPPE
67335Atto 488DOPE
40692Atto 532 DOPE
42208Atto 550 DOPE
05676Atto594 DOPE
90077Atto 633 DOPE
42247Atto 647N DOPE
54368Atto 488 DMPE
06713Atto 532 DMPE
42971Atto 550 DMPE
72567Atto 594 DMPE
55284Atto 633 DMPE
89522Atto 647 DMPE
51028Atto 488 PPE
94092Atto 532 PPE
78998Atto 550 PPE
19504Atto 594 PPE
56530Atto 633 PPE
91327Atto 647N PPE
Materials
Sorry, an unexpected error has occurred
Network error: Failed to fetch
References
1.
Neuweiler H, Schulz A, Vaiana AC, Smith JC, Kaul S, Wolfrum J, Sauer M. 2002. Detection of Individual p53-Autoantibodies by Using Quenched Peptide-Based Molecular Probes. Angew. Chem. Int. Ed.. 41(24):4769-4773. https://doi.org/10.1002/anie.200290044
2.
Sauer M, Zander C, Müller R, Ullrich B, Drexhage K, Kaul S, Wolfrum J. 1997. Detection and identification of individual antigen molecules in human serum with pulsed semiconductor lasers. Applied Physics B: Lasers and Optics. 65(3):427-431. https://doi.org/10.1007/s003400050292
3.
Sauer M, Zander C, Müller R, Ullrich B, Drexhage K, Kaul S, Wolfrum J. 1997. Detection and identification of individual antigen molecules in human serum with pulsed semiconductor lasers. Applied Physics B: Lasers and Optics. 65(3):427-431. https://doi.org/10.1007/s003400050292
4.
Widengren J, Schweinberger E, Berger S, Seidel CAM. 2001. Two New Concepts to Measure Fluorescence Resonance Energy Transfer via Fluorescence Correlation Spectroscopy:  Theory and Experimental Realizations. J. Phys. Chem. A. 105(28):6851-6866. https://doi.org/10.1021/jp010301a
5.
Buschmann V, Weston KD, Sauer M. 2003. Spectroscopic Study and Evaluation of Red-Absorbing Fluorescent Dyes. Bioconjugate Chem.. 14(1):195-204. https://doi.org/10.1021/bc025600x
6.
Widengren J, Schwille P. 2000. Characterization of Photoinduced Isomerization and Back-Isomerization of the Cyanine Dye Cy5 by Fluorescence Correlation Spectroscopy. J. Phys. Chem. A. 104(27):6416-6428. https://doi.org/10.1021/jp000059s