Merck
HomeProtein ExpressionHistamine Receptors
Histamine Receptors
The biogenic amine histamine plays an important role in a variety of pathophysiological conditions. In peripheral tissues, histamine is mainly stored in mast cells and basophils. In allergic conditions, histamine is released from these cells and is responsible for several of the well known symptoms of allergic conditions of the skin and airways. In the gastric mucosa, gastrin-induced histamine release fulfills an important physiological role by stimulating parietal cells to secrete gastric acid. In the CNS, histamine is synthesized in specific neurons that are localized in the tubero-mammillary nucleus of the posterior hypothalamus. These neurons project to all major brain areas and are involved in a variety of important physiological functions, including the regulation of the sleep-wake cycle, food intake, cardiovascular control, regulation of the hypothalamic pituitary adrenal-axis (HPA-axis), learning and memory.
Histamine exerts its action via at least four distinct receptor subtypes. Molecular biological approaches have shown that all histamine receptors belong to the large family of G protein-coupled receptors. The gene encoding the Hreceptor has only recently been cloned. In contrast to the H1 and H2 receptor gene, the H3 receptor gene contains intronic sequences, leading to the identification of various H3 receptor isoforms following alternative splicing of the introns. The isoforms show distinct expression patterns and signal transduction mechanisms. Using the H3 receptor sequence, a new histamine (H4) receptor was identified 'in silico'. This receptor shows the strongest homology to the H3 receptor and also recognizes histamine with high affinity.
The H1 receptor couples mainly to Gq/11 thereby stimulating phospholipase C, whereas the H2 receptor interacts with Gs to activate adenylyl cyclase. The histamine H3 and H4 receptors couple to Gi proteins to inhibit adenylyl cyclase, and to stimulate MAPK.
Many potent and selective antagonists for the H1 and H2 receptors have been developed as successful anti-allergic or anti-ulcer drugs. Selective agonists are currently also available as pharmacological tools. The H3 receptor was originally described as an autoreceptor, inhibiting the release of histamine from histaminergic neurons in brain. Recently, it was shown that this inhibitory effect is due to constitutive activity of the H3 receptor. Evidence suggests that the H3 receptor regulates the release of several important neurotransmitters (e.g. acetylcholine, dopamine, GABA, norepinephrine, serotonin), both in the peripheral and central nervous systems. Highly potent and selective agonists and antagonists have been developed for the H3 receptor. These ligands are useful pharmacological tools and are currently being assessed for their clinical potential in allergy, inflammatory disorders, attention deficit hyperactivity disorder, Alzheimer's disease and obesity.
The H4 receptor is highly expressed in peripheral blood leukocytes and intestinal tissue, making this receptor a potentially interesting target in allergic and inflammatory diseases. The receptor shows high affinity for several H3 receptor ligands (both agonists and antagonists), but selective antagonists and agonists have been described.
Because of the availability of many potent and subtype selective ligands for histamine receptor subtypes, good radioligands are available. For the H1 receptor, the antagonist [3H]-mepyramine has been successfully used in many preparations, although binding to cytochrome P450 isoenzymes may mask H1 receptor binding. For the H2 receptor, the antagonist [125I]-iodoaminopotentidine has recently been developed as a high affinity radioligand. As for the H1 receptor, an agonist radioligand is lacking. In contrast, agonist and antagonist radioligands are available for the H3 receptor. Initially, the agonists Nα-methylhistamine and (R)-α-methyl-histamine were developed as tritiated radioligands. Both ligands show high affinity labeling of the H3 receptor with almost no non-specific binding. Originally described as an antagonist, iodoproxyfan acts as a partial agonist in some H3 receptor models. The iodonated ligand [125I]-iodoproxyfan can therefore also be used as an agonist radioligand. [125I]-Iodophenpropit, [3H]-GR168320, [3H]-clobenpropit and can all be used as H3 receptor antagonist radioligands. For the H4 receptor, [3H]-histamine or [3H]-JNJ7777120 can be used as respectively agonist or antagonist radiolabel.
The Table below contains accepted modulators and additional information. For a list of additional products, see the "Similar Products" section below.
Currently Accepted NameH1H2H3H4
Structural Information487 aa (human)359 aa (human)445 aa (human)390 aa (human)
Subtype Selective
Agonists
2-((3-Trifluoromethyl)-
phenyl)histaminea (T4951)
N-Methylhistaprodifen
Amthamine (A4730)
Immethridine (I1909)
N-Methylimmepip
4-Methylhistamine
Clobenpropit (C209)
Clozapine (C6305)
Subtype Selective
Antagonists
(+)-Chlorpheniramine (C4915)
Pyrilamine (Mepyramine) (P5514)
Triprolidine (T6764)
Cimetidine (C4522)
Ranitidine (R101)
Tiotidine
Clobenpropitb (C209)
Ciproxifanc (C6848)
JNJ7777120 (J3770)
Receptor Selective
Agonists
Nα-Methylhistamine (H5914)
Histamine (H7250)
Nα-Methylhistamine (H5914)
Histamine (H7250)
Histamine (H7250)
Nα-Methylhistamine (H5914)
R-α-Methylhistamine (H128)
Immepip (I4034)
Imetit (I135)
Histamine (H7250)
Nα-Methylhistamine (H5914)
R-α-Methylhistamine (H128)         
Immepip (I4034)
Imetit (I135)
Receptor Selective
Antagonists
Not KnownNot KnownThioperamide (T123)
Thioperamide (T123)
Signal Transduction MechanismsGq/11 (increase IP3/DAG)Gs (increase cAMP)Gi (decrease cAMP)Gi (decrease cAMP)
Radioligands of Choice[3H]-Pyrilamine (Mepyramine)[125I]-Iodoaminopotentidine  
[3H]-Nα-Methylhistamine
[125I]-Iodophenpropit
[3H]-Histamine
[3H]-JNJ7777120
Tissue ExpressionLung, vasculature, brainHeart, stomach, brainNeurons (CNS, PNS)Mast cells, eosinophils
Physiological FunctionSmooth muscle contraction,
food intake, sleep-wakefulness
Gastric acid secretionRegulation sleep,
food intake, cognition
Chemotaxis
Disease RelevanceAllergic conditionsGastric ulcersCognitive impairment, obesityInflammation
Footnotes
a) 2-((3-Trifluoromethyl)phenyl)histamine and N-methylhistaprodifen are full agonists in the guinea-pig ileum, the standard assay system for H1 receptors. In other systems, the compound may act as a partial agonists, as is the case for many histaminergic agonists, e.g. impromidine and dimaprit at the H2 receptor.
b) In vitro, clobenpropit is one of the most potent H3 receptor antagonists known at the present time (pA2 = 9.9). It also displays partial H4 agonist activity.
c) Ciproxifan demonstrates good CNS penetration and is 100 times more potent in vivo than clobenpropit. Its activity at human H3 receptors is 100-fold lower than at rat H3 receptors and in the same order as at human α2 receptors.
Similar Products
Sorry, an unexpected error has occurred
Network error: Failed to fetch
References
1.
DEESCH I, THURMOND R, JONGEJAN A, LEURS R. 2005. The histamine H receptor as a new therapeutic target for inflammation. Trends in Pharmacological Sciences. https://doi.org/10.1016/j.tips.2005.07.002
2.
Drutel G, Peitsaro N, Karlstedt K, Wieland K, Smit MJ, Timmerman H, Panula P, Leurs R. 2001. Identification of Rat H3Receptor Isoforms with Different Brain Expression and Signaling Properties. Mol Pharmacol. 59(1):1-8. https://doi.org/10.1124/mol.59.1.1
3.
Hill S, Ganellin C, Timmerman H, Schwartz J, Shankley N, Young J, Schunack W, Levi R, Haas H. 1997. International Union of Pharmacology. XIII. Classification of histamine receptorss Pharmacological reviews . 49(3):253-278.
4.
Hu W, Chen Z. 2012. Role of Histamine and Its Receptors in Cerebral Ischemia. ACS Chem. Neurosci.. 3(4):238-247. https://doi.org/10.1021/cn200126p
5.
Leurs R, Bakker RA, Timmerman H, de Esch IJP. 2005. The histamine H3 receptor: from gene cloning to H3 receptor drugs. Nat Rev Drug Discov. 4(2):107-120. https://doi.org/10.1038/nrd1631
6.
Lim HD, van Rijn RM, Ling P, Bakker RA, Thurmond RL, Leurs R. 2005. Evaluation of Histamine H1-, H2-, and H3-Receptor Ligands at the Human Histamine H4 Receptor: Identification of 4-Methylhistamine as the First Potent and Selective H4 Receptor Agonist. J Pharmacol Exp Ther. 314(3):1310-1321. https://doi.org/10.1124/jpet.105.087965
7.
Morisset S, Rouleau A, Ligneau X, Gbahou F, Tardivel-Lacombe J, Stark H, Schunack W, Ganellin CR, Arrang J. 2000. High constitutive activity of native H3 receptors regulates histamine neurons in brain. Nature. 408(6814):860-864. https://doi.org/10.1038/35048583
8.
Nijmeijer S. 2012. Molecular pharmacology of histamine H4 receptors. Front Biosci. 17(7):2089. https://doi.org/10.2741/4039
9.
Oda T, Morikawa N, Saito Y, Masuho Y, Matsumoto S. 2000. Molecular Cloning and Characterization of a Novel Type of Histamine Receptor Preferentially Expressed in Leukocytes. J. Biol. Chem.. 275(47):36781-36786. https://doi.org/10.1074/jbc.m006480200
10.
O?Mahony L, Akdis M, Akdis CA. 2011. Regulation of the immune response and inflammation by histamine and histamine receptors. Journal of Allergy and Clinical Immunology. 128(6):1153-1162. https://doi.org/10.1016/j.jaci.2011.06.051
11.
Passani MB, Lin J, Hancock A, Crochet S, Blandina P. 2004. The histamine H3 receptor as a novel therapeutic target for cognitive and sleep disorders. Trends in Pharmacological Sciences. 25(12):618-625. https://doi.org/10.1016/j.tips.2004.10.003
12.
Passani MB, Blandina P. 2011. Histamine receptors in the CNS as targets for therapeutic intervention. Trends in Pharmacological Sciences. 32(4):242-249. https://doi.org/10.1016/j.tips.2011.01.003
13.
Schwartz JC, Arrang JM, Garbarg M, Pollard H, Ruat M. 1991. Histaminergic transmission in the mammalian brain. Physiological Reviews. 71(1):1-51. https://doi.org/10.1152/physrev.1991.71.1.1
14.
Thurmond RL, Desai PJ, Dunford PJ, Fung-Leung W, Hofstra CL, Jiang W, Nguyen S, Riley JP, Sun S, Williams KN, et al. 2004. A Potent and Selective Histamine H4 Receptor Antagonist with Anti-Inflammatory Properties. J Pharmacol Exp Ther. 309(1):404-413. https://doi.org/10.1124/jpet.103.061754
15.
Walter M, Kottke T, Stark H. 2011. The histamine H4 receptor: Targeting inflammatory disorders. European Journal of Pharmacology. 668(1-2):1-5. https://doi.org/10.1016/j.ejphar.2011.06.029
16.
Yamaura K, Shigemori A, Suwa E, Ueno K. 2013. Expression of the histamine H4 receptor in dermal and articular tissues. Life Sciences. 92(2):108-113. https://doi.org/10.1016/j.lfs.2012.10.030
17.
Wolff M. 1997 . Burger's Medicinal Chemistry an Drug Discovery . fifth edition . New York: John Wiley & Sons pp. 495-559.
18.
Zingel V, Leschke C, Schunack W. 1995. Developments in histamine H1-receptor agonists.49-85. https://doi.org/10.1007/978-3-0348-7161-7_2