Merck
HPLC Analysis of Phensuximide Enantiomers on Astec® CYCLOBOND I 2000 DMP
HPLC Analysis of Phensuximide Enantiomers on Astec® CYCLOBOND I 2000 DMP application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Astec® CYCLOBOND I 2000 DMP Chiral HPLC Column
5 μm particle size, L × I.D. 25 cm × 4.6 mm
CONDITIONS
column
CYCLOBOND I 2000 DMP, 25 cm x 4.6 mm I.D., 5 μm particles (20724AST)
mobile phase
[A] isopropanol; [B] hexane; (80:20, A:B)
flow rate
1 mL/min
column temp.
22 °C
detector
UV, 254 nm
injection
10 μL
sample
4 mg/mL in methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
null