HPLC Analysis of Ciprofibrate Enantiomers on Astec® CYCLOBOND I 2000
![HPLC Analysis of Ciprofibrate Enantiomers on Astec® CYCLOBOND I 2000 application for HPLC](/deepweb/assets/sigmaaldrich/product/images/421/343/d16b8782-77c9-45a8-a4b2-626200225790/800/d16b8782-77c9-45a8-a4b2-626200225790.jpg)
CONDITIONS
column
CYCLOBOND I 2000, 25 cm x 4.6 mm I.D., 5 μm particles (20024AST)
mobile phase
[A] acetonitrile; [B] methanol; [C] acetic acid; [D] triethylamine; (90:10:0.3:0.2, A:B:C:D)
flow rate
1 mL/min
column temp.
ambient
detector
UV, 230 nm
injection
1 μL
sample
5 mg/mL in methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
null