GC Analysis of δ-Decanolactone Enantiomers on Astec® CHIRALDEX™ G-TA
![GC Analysis of δ-Decanolactone Enantiomers on Astec® CHIRALDEX™ G-TA suitable for GC](/deepweb/assets/sigmaaldrich/product/images/223/699/4b624df7-a9d5-4ad3-8e3a-44a8b02594b6/800/4b624df7-a9d5-4ad3-8e3a-44a8b02594b6.jpg)
CONDITIONS
column
Astec CHIRALDEX G-TA, 30 m x 0.25 mm I.D., 0.12 μm (73033AST)
oven
helium, 24 psi
inj. temp.
250 °C
detector
FID, 250 °C
carrier gas
helium, 24 psi
sample
peaks 1 & 2: δ-decanolactone enantiomers
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIRALDEX is a trademark of Sigma-Aldrich Co. LLC