Merck
  • Eriodictyol Attenuates Myocardial Ischemia-Reperfusion Injury through the Activation of JAK2.
Eriodictyol Attenuates Myocardial Ischemia-Reperfusion Injury through the Activation of JAK2.
Frontiers in pharmacology (2018-02-15)
Defang Li, Ning Lu, Jichun Han, Xiaoyu Chen, Wenjin Hao, Wenjuan Xu, Xiaona Liu, Lei Ye, Qiusheng Zheng
ABSTRACT
Myocardial ischemia-reperfusion (I/R) injury remains the leading risk factor of disability and mortality worldwide. In this study, the myocardial protective effect of eriodictyol (EDT) and the underlying mechanism in an