- HOXA3 promotes tumor growth of human colon cancer through activating EGFR/Ras/Raf/MEK/ERK signaling pathway.
HOXA3 promotes tumor growth of human colon cancer through activating EGFR/Ras/Raf/MEK/ERK signaling pathway.
Homeobox A3 (HOXA3), one of HOX transcription factors, regulates gene expression during embryonic development. HOXA3 expression has been reported to be associated with several cancers; however, its role in colon cancer and underlying mechanism are still unclear. The expression of HOXA3 in 232 paired of human colon tumor and adjacent non-tumorous tissues were measured by qPCR. The relationship between HOXA3 expression and clinical outcomes were analyzed by Kaplan-Meier survival curves analysis. Human colon cancer cell lines HT29 and HTC116 were transfected with HOXA3 siRNA, or HOXA3 expressing vector, and then cell proliferation and apoptosis were assessed, respectively. Western blot was performed to detect the activation of EGFR/Ras/Raf/MEK/ERK signaling pathway. Moreover, HOXA3-overexpressing and HOXA3-suppressing HT29 cells were subcutaneous injected into nod mice to confirm the regulation of HOXA3 on EGFR/Ras/Raf/MEK/ERK signaling in regulating tumor growth. HOXA3 was upregulated in colon tumor tissues and cell lines, and upregulated expression of HOXA3 was associated with low survival rate. Knockdown of HOXA3 suppressed cell viability and clone formation, while induced cell apoptosis. HOXA3 knockdown could not induce the increase of cell apoptosis on the condition of EGFR overexpression. In vivo xenograft studies, HOXA3-suppressing cells showed less tumorigenic. Moreover, HOXA3 knockdown suppressed the activation of EGFR/Ras/Raf/MEK/ERK signaling pathway. To conclude, this study indicated that HOXA3 might act as a promoter of human colon cancer formation by regulating EGFR/Ras/Raf/MEK/ERK signaling pathway. HOXA3 might be a potential therapeutic target for the treatment of colon cancer.