Merck
  • Lysine acetylome profiling uncovers novel histone deacetylase substrate proteins in
Lysine acetylome profiling uncovers novel histone deacetylase substrate proteins in
Molecular systems biology (2017-10-25)
Markus Hartl, Magdalena Füßl, Paul J Boersema, Jan-Oliver Jost, Katharina Kramer, Ahmet Bakirbas, Julia Sindlinger, Magdalena Plöchinger, Dario Leister, Glen Uhrig, Greg Bg Moorhead, Jürgen Cox, Michael E Salvucci, Dirk Schwarzer, Matthias Mann, Iris Finkemeier
ABSTRACT
Histone deacetylases have central functions in regulating stress defenses and development in plants. However, the knowledge about the deacetylase functions is largely limited to histones, although these enzymes were found in diverse subcellular compartments. In this study, we determined the proteome-wide signatures of the RPD3/HDA1 class of histone deacetylases in
Sigma-Aldrich
Tetramethylrhodamine methyl ester perchlorate, ≥95%