Merck
  • TLE1 inhibits anoikis and promotes tumorigenicity in human lung cancer cells through ZEB1-mediated E-cadherin repression.
TLE1 inhibits anoikis and promotes tumorigenicity in human lung cancer cells through ZEB1-mediated E-cadherin repression.
Oncotarget (2017-10-27)
Xin Yao, Tri Pham, Brandi Temple, Selena Gray, Cornita Cannon, Camry Hardy, Kamari Fletcher, Shubha Kale Ireland, Ahamed Hossain, Renwei Chen, Asim B Abdel-Mageed, Hector Biliran
ABSTRACT
The Transducin-like enhancer of split 1 (TLE1) corepressor protein is overexpressed in human lung tumors and is a putative lung-specific oncogene. However, the molecular mechanism underlying its oncogenic function remains to be delineated. Here, we report an important role of TLE1 in promoting lung tumorigenesis by a mechanism involving induction of anoikis resistance. Using the human lung adenocarcinoma A549 and immortalized bronchial epithelial BEAS-2B cell lines, we observed that TLE1 inhibits anoikis through transcriptional repression of E-cadherin gene. In support of E-cadherin as a downstream target of TLE1 to block anoikis, forced expression of E-cadherin attenuated TLE1-induced anoikis resistance while E-cadherin downregulation decreased the anoikis sensitivity of TLE1 knockdown cells. Furthermore, we determined that E-cadherin expression is transcriptionally induced upon loss of cell attachment and functions as an effector of anoikis. Loss of E-cadherin via the siRNA strategy or exogenous TLE1 expression was sufficient to attenuate anoikis in A549 and BEAS-2B cells. Importantly, we demonstrated that the ZEB1 transcriptional factor is required for TLE1-mediated E-cadherin repression and anoikis resistance. ZEB1 interacted with and recruited the TLE1 to the E-cadherin promoter to impose histone deacetylation and gene silencing. In vivo, TLE1 strongly promoted tumorigenicity of A549 cells in a ZEB1-dependent manner. Underscoring its role in anoikis insensitivity of lung cancer cells, the TLE1-mediated E-cadherin repression was negatively regulated by the tumor suppressor Bcl-2 inhibitor of transcription 1 (Bit1) to effect anoikis. These findings identify the ZEB1/TLE1/E-cadherin transcriptional mechanism as a novel pathway that promotes anoikis resistance and oncogenicity of lung cancer cells.
Sigma-Aldrich
MISSION® esiRNA, targeting human ZEB1
Sigma-Aldrich
MISSION® esiRNA, targeting human TLE1