Merck
  • Stimulation of CCL2 (MCP-1) and CCL2 mRNA by substance P in LAD2 human mast cells.
Stimulation of CCL2 (MCP-1) and CCL2 mRNA by substance P in LAD2 human mast cells.
Translational research : the journal of laboratory and clinical medicine (2009-06-16)
Maria Luisa Castellani, Jacopo Vecchiet, Vincenzo Salini, Pio Conti, Theoharis C Theoharides, Auro Caraffa, Pierluigi Antinolfi, Stefano Teté, Cristian Ciampoli, Chiara Cuccurullo, Giuliano Cerulli, Mario Felaco, Paolo Boscolo
ABSTRACT
Chemokines are cytokines with chemotactic properties on inflammatory cells and other cell types. Chemokine (C-C motif) ligand 2 (CCL2), which is also called monocyte chemotactic protein 1 (MCP-1), is a potent chemotactic molecule that attracts lymphocytes, monocytes, mast cells, and memory T cells, but not neutrophils. CCL2/MCP-1 represents a link between the activation of monocytes, lymphocytes, basophils, mast cells, and eosinophils in inflammatory disorders, such as the late-phase allergic reaction. This C-C chemokine also plays a role in regulating Th-cell cytokine production and leukocyte trafficking. Laboratory of allergic diseases (LAD) cells is the first reported human mast cell line that closely resembles a primary culture of CD34+-derived human mast cells. These cells were cultured in vitro and treated with different concentrations of substance P (SP) for the production of CCL2/MCP-1. We used calcium ionophore as a positive control for stimulating transcription and translation of CCL2/MCP-1. The stimulation of SP on CCL2/MCP-1 was statistically significant (P < 0.05) compared with the control (untreated cells). In this study, we determined the expression and secretion of CCL2/MCP-1 from SP-activated LAD2 human mast cells in vitro. The levels of CCL2/MCP-1 from SP-activated LAD2 human mast cells were higher at 10 microM and at 18 h incubation compared with controls. This effect was also revealed on CCL2/MCP-1 messenger RNA (mRNA) expression, as determined by reverse transcriptase polymerase chain reaction (RT-PCR) analysis. Our data suggest that SP is an important neurotransmitter that can stimulate the chemokine CCL2, which plays a fundamental role in inflammation by recruiting inflammatory cells to specific cites.
Sigma-Aldrich
Substance P acetate salt hydrate, ≥95% (HPLC), powder