Merck
Advancing polyethylene properties by incorporating NO
Dalton transactions (Cambridge, England : 2003) (2017-05-16)
Qaiser Mahmood, Yanning Zeng, Xinxin Wang, Yang Sun, Wen-Hua Sun
ABSTRACT
A new family of nickel halides (bromides Ni1-Ni5 and chlorides Ni6-Ni10) ligated by 1-(2,6-dibenzhydryl-4-nitrophenylimino)-2-(arylimino)acenaphthylene (Aryl = 2,6-Me