Merck
  • Rapid multiresidue determination of bisphenol analogues in soil with on-line derivatization.
Rapid multiresidue determination of bisphenol analogues in soil with on-line derivatization.
Analytical and bioanalytical chemistry (2017-06-03)
Rosa Ana Pérez, Beatriz Albero, Macarena Férriz, José Luis Tadeo
ABSTRACT
Bisphenol analogues are compounds extensively used which have been potentially linked to adverse health effects. Nevertheless, few studies reported the analysis of compounds, other than bisphenol A, in environmental solid samples and none in soil samples. In this study, a rapid and sensitive analytical method is presented for the simultaneous determination of 13 bisphenols in soil samples. The method combines ultrasonic-assisted extraction of samples placed in small columns and GC-MS/MS analysis. Manual and on-line derivatizations were compared and results showed that significant higher chromatographic responses were achieved with on-line derivatization. Different parameters such as the quantity of derivatization agent, the extraction solvent, or the extraction time were assayed. The detection limits for all target bisphenols ranged from 0.04 to 0.27 ng g
Product Description
Supelco
Tetrachlorobisphenol A, analytical standard
Sigma-Aldrich
Trifluoroacetamide, 97%