Merck
Dysregulation of type 2 innate lymphoid cells and T
The Journal of allergy and clinical immunology (2016-06-19)
Katrien C De Grove, Sharen Provoost, Rudi W Hendriks, Andrew N J McKenzie, Leen J M Seys, Smitha Kumar, Tania Maes, Guy G Brusselle, Guy F Joos
ABSTRACT
Although the prominent role of T We sought to investigate the relative contribution of ILC2 and adaptive T Wild-type, Gata-3 Concomitant DEP+HDM exposure significantly enhanced allergic airway inflammation, as characterized by increased airway eosinophilia, goblet cell metaplasia, accumulation of ILC2s and T These data indicate that dysregulation of ILC2s and T
Sigma-Aldrich
Human IL-33 ELISA Kit, for serum, plasma, cell culture supernatant and urine