Merck
  • Interleukin-33 produced by M2 macrophages and other immune cells contributes to Th2 immune reaction of IgG4-related disease.
Interleukin-33 produced by M2 macrophages and other immune cells contributes to Th2 immune reaction of IgG4-related disease.
Scientific reports (2017-02-17)
Sachiko Furukawa, Masafumi Moriyama, Kensuke Miyake, Hitoshi Nakashima, Akihiko Tanaka, Takashi Maehara, Mana Iizuka-Koga, Hiroto Tsuboi, Jun-Nosuke Hayashida, Noriko Ishiguro, Masaki Yamauchi, Takayuki Sumida, Seiji Nakamura
ABSTRACT
IgG4-related disease (IgG4-RD) is characterized by elevated serum IgG4 and marked infiltration of IgG4-positive cells in multiple organs. Interleukin-33 (IL-33) is a recently described cytokine that is secreted by damaged epithelial cells, macrophages, and dendritic cells, and potently activates helper T type 2 (Th2) immune responses, which have been suggested to play a major role in IgG4 production of IgG4-RD. Here, we assessed the expression of IL-33 and related molecules in the salivary glands (SGs) of patients with IgG4-RD versus that in patients with Sjögren's syndrome (SS) and controls. Expression of IL-33 and its receptor (ST2) was strongly detected around ectopic germinal centers (GCs) in the SGs from patients with IgG4-RD, whereas IL-33 was expressed only in epithelial cells in patients with SS and controls. Moreover, IL-33 and CD68
Sigma-Aldrich
Human IL-33 ELISA Kit, for serum, plasma, cell culture supernatant and urine