Merck
  • A Rhodamine-based Dual Chemosensor for the Simultaneous Detection of Fe3+ and Cu2.
A Rhodamine-based Dual Chemosensor for the Simultaneous Detection of Fe3+ and Cu2.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry (2016-11-11)
Tao Lan, Feng-He Wang, Xing-Jun Xi, Chun-Wen Cheng, Wu Lei, Ming-Zhu Xia, Feng-Yun Wang
ABSTRACT
A Rhodamine-based dual chemosensor L1 for simultaneously detecting Fe3+ and Cu2+ was designed and synthesized. The spectroscopic properties of L1 were analyzed, and its recognition mechanism was speculated. We found that the addition of Fe3+ induced a great fluorescence enhancement, while Cu2+ induced a strong UV-Vis absorption enhancement. The results revealed that L1 was highly selective for recognizing Fe3+ and Cu2+ in UV-Vis spectroscopy in CH3OH-H2O (1/1, v/v, pH 7.2) with the interference of other metal ions. A good linear relationship between the fluorescence intensities of L1 and the concentration of Fe3+, as well as the UV-Vis absorption intensities of L1 and the concentration of Cu2+ was observed, respectively. The detection limit was 9.2 × 10-8 M (5.5 μg/L) for Fe3+ and 3.8 × 10-8 M (2.4 μg/L) for Cu2+, respectively. The detection capacity for targeted metal ions of Fe3+ and Cu2+ were studied, which are less than 5 min. Job's plot method for L1 with Fe3+ and ESI-MS for L1 with Cu2+ indicated a 1:1 stoichiometry in the complex. The results may provide an effective strategy for the design of new dual chemosensors for the rapid detection of targeted metal ions.
Sigma-Aldrich
2-Thiophenecarboxaldehyde, 98%