Merck
  • Catalytic enantioselective hetero-Diels-Alder reactions of an azo compound.
Catalytic enantioselective hetero-Diels-Alder reactions of an azo compound.
Journal of the American Chemical Society (2006-12-21)
Masanori Kawasaki, Hisashi Yamamoto
ABSTRACT
This communication describes studies in which an azo hetero-Diels-Alder adduct was furnished in high regio- and enantioselectivity using azopyridine as a reagent and silver as a catalyst. The obtained hetero-Diels-Alder adduct was easily converted to the corresponding chiral 1,4-diamino alcohol.
Sigma-Aldrich
(R)-(+)-2,2′-Bis(diphenylphosphino)-1,1′-binaphthalene, 97%