Merck
  • Induction of DARPP-32 by brain-derived neurotrophic factor in striatal neurons in vitro is modified by histone deacetylase inhibitors and Nab2.
Induction of DARPP-32 by brain-derived neurotrophic factor in striatal neurons in vitro is modified by histone deacetylase inhibitors and Nab2.
PloS one (2013-11-10)
Samira Chandwani, Serene Keilani, Maitane Ortiz-Virumbrales, Andrika Morant, Steve Bezdecny, Michelle E Ehrlich
ABSTRACT
Neurotrophins and modifiers of chromatin acetylation and deacetylation participate in regulation of transcription during neuronal maturation and maintenance. The striatal medium spiny neuron is supported by cortically-derived brain derived neurotrophic factor and is the most vulnerable neuron in Huntington's disease, in which growth factor and histone deacetylase activity are both disrupted. We examined the ability of three histone deacetylase inhibitors, trichostatin A, valproic acid and Compound 4 b, alone and combined with brain derived neurotrophic factor (BDNF), to promote phenotypic maturation of striatal medium spiny neurons in vitro. Exposure of these neurons to each of the three compounds led to an increase in overall histone H3 and H4 acetylation, dopamine and cyclic AMP-regulated phosphoprotein, 32 kDa (DARPP-32) mRNA and protein, and mRNA levels of other markers of medium spiny neuron maturation. We were, however, unable to prove that HDAC inhibitors directly lead to remodeling of Ppp1r1b chromatin. In addition, induction of DARPP-32 by brain-derived neurotrophic factor was inhibited by histone deacetylase inhibitors. Although BDNF-induced increases in pTrkB, pAkt, pERK and Egr-1 were unchanged by combined application with VPA, the increase in DARPP-32 was relatively diminished. Strikingly, the NGF1A-binding protein, Nab2, was induced by BDNF, but not in the presence of VPA or TSA. Gel shift analysis showed that α-Nab2 super-shifted a band that is more prominent with extract derived from BDNF-treated neurons than with extracts from cultures treated with VPA alone or VPA plus BDNF. In addition, overexpression of Nab2 induced DARPP-32. We conclude that histone deacetylase inhibitors inhibit the induction of Nab2 by BDNF, and thereby the relative induction of DARPP-32.