Merck
  • In vivo pharmacology and antidiarrheal efficacy of a thiazolidinone CFTR inhibitor in rodents.
In vivo pharmacology and antidiarrheal efficacy of a thiazolidinone CFTR inhibitor in rodents.
Journal of pharmaceutical sciences (2005-03-12)
N D Sonawane, Chatchai Muanprasat, Ray Nagatani, Yuanlin Song, A S Verkman
ABSTRACT
A small-molecule inhibitor of the cystic fibrosis transmembrane conductance regulator (CFTR), 3-[(3-trifluoromethyl)phenyl]-5-[(4-carboxyphenyl)methylene]-2-thioxo-4-thiazolidinone (CFTR(inh)-172), reduces enterotoxin-induced intestinal fluid secretion in rodents. Here, we study CFTR(inh)-172 pharmacology and antidiarrheal efficacy in rodents using (14)C-labeled CFTR(inh)-172, liquid chromatography/mass spectrometry, and a closed intestinal loop model of fluid secretion. CFTR(inh)-172 was cleared primarily by renal glomerular filtration without chemical modification. CFTR(inh)-172 accumulated in liver within 5 min after intravenous infusion in mice, and was concentrated fivefold in bile over blood. At 30-240 min, CFTR(inh)-172 was found mainly in liver, intestine, and kidney, with little detectable in the brain, heart, skeletal muscle, or lung. Pharmacokinetic analysis in rats following intravenous bolus infusion showed a distribution volume of 770 mL with redistribution and elimination half-times of 0.14 h and 10.3 h, respectively. CFTR(inh)-172 was stable in hepatic microsomes. Closed-loop studies in mice indicated that a single intraperitoneal injection of 20 microg CFTR(inh)-172 inhibited fluid accumulation at 6 h after cholera toxin by >90% in duodenum and jejunum, approximately 60% in ileum and <10% in colon. No toxicity was seen after high-dose CFTR(inh)-172 administration (3 mg/kg/day in two daily doses) in mice over the first 6 weeks of life. The metabolic stability, enterohepatic recirculation, slow renal elimination, and intestinal accumulation of CFTR(inh)-172 account for its efficacy as an antidiarrheal.
Sigma-Aldrich
CFTR(inh)-172, ≥98% (HPLC), powder