Merck
Get up to 22% off for Pi Day until 3/26.Save Now
  • The MEKK1 PHD ubiquitinates TAB1 to activate MAPKs in response to cytokines.
The MEKK1 PHD ubiquitinates TAB1 to activate MAPKs in response to cytokines.
The EMBO journal (2014-09-28)
Nikolaos Charlaftis, Tesha Suddason, Xuefeng Wu, Saba Anwar, Michael Karin, Ewen Gallagher
ABSTRACT
Unlike the other MAP3Ks, MEKK1 (encoded by Map3k1) contains a PHD motif. To understand the role of this motif, we have created a knockin mutant of mouse Map3k1 (Map3k1(m) (PHD)) with an inactive PHD motif. Map3k1(m) (PHD) ES cells demonstrate that the MEKK1 PHD controls p38 and JNK activation during TGF-β, EGF and microtubule disruption signalling, but does not affect MAPK responses to hyperosmotic stress. Protein microarray profiling identified the adaptor TAB1 as a PHD substrate, and TGF-β- or EGF-stimulated Map3k1(m) (PHD) ES cells exhibit defective non-canonical ubiquitination of MEKK1 and TAB1. The MEKK1 PHD binds and mediates the transfer of Lys63-linked poly-Ub, using the conjugating enzyme UBE2N, onto TAB1 to regulate TAK1 and MAPK activation by TGF-β and EGF. Both the MEKK1 PHD and TAB1 are critical for ES-cell differentiation and tumourigenesis. Map3k1(m) (PHD) (/+) mice exhibit aberrant cardiac tissue, B-cell development, testis and T-cell signalling.
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, ascites fluid, clone B-5-1-2
Sigma-Aldrich
Anti-MAP Kinase (ERK-1, ERK-2) antibody produced in rabbit, whole antiserum
Sigma-Aldrich
Monoclonal Anti-T7 tag antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-MAP Kinase, Activated (Diphosphorylated ERK-1&2) antibody produced in mouse, clone MAPK-YT, ascites fluid
Millipore
ANTI-FLAG® antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution