Merck
Get up to 22% off for Pi Day until 3/26.Save Now
  • Diamino Telechelic Polybutadienes for Solventless Styrene-butadiene-styrene (SBS) Triblock Copolymer Formation.
Diamino Telechelic Polybutadienes for Solventless Styrene-butadiene-styrene (SBS) Triblock Copolymer Formation.
Polymer (2009-11-13)
Shengxiang Ji, Thomas R Hoye, Christopher W Macosko
ABSTRACT
High molecular weight, high functionality diamino telechelic polybutadienes (TPBs) were synthesized by ring-opening metathesis polymerization (ROMP) of 1,5-cyclooctadiene (COD) in the presence of a chain transfer agent, 1,8-dicyano-4-octene, followed by lithium aluminum hydride reduction. Melt coupling of diamino TPB with anhydride-terminated polystyrene (PS-anh) resulted in the formation of styrene-butadiene-styrene (SBS) triblock copolymers; ca. 80% maximum conversion of PS-anh was achieved within 30 seconds. The results from SAXS, TEM, and rheological measurements of the coupling products confirmed the formation of SBS triblock copolymers having lamellar morphology. A fluororesent-labeled PS-anh was used to study the coupling kinetics by diluting the reactants by the addition of non-functional PS.