Merck
  • Netrin-1 promotes cell migration and invasion by down-regulation of BVES expression in human hepatocellular carcinoma.
Netrin-1 promotes cell migration and invasion by down-regulation of BVES expression in human hepatocellular carcinoma.
American journal of cancer research (2015-06-24)
Ping Han, Yu Fu, Jingmei Liu, Yunwu Wang, Jiayi He, Jin Gong, Mengke Li, Qinghai Tan, Dongxiao Li, Yixing Luo, Jian Han, Jiqiao Liu, Wei Tu, Ying Wang, Dean Tian, Wei Yan
ABSTRACT
The axon guidance cues netrin-1 has been reported to be associated with cancer progression in various types of human cancers. However, the underlying molecular mechanism of netrin-1-mediated metastasis remains obscure. In this study, we found that overexpression of netrin-1 promoted HCC cell migration and invasion as determined by transwell assay and 3D cell culture assay. However, netrin-1 knockdown inhibited these processes. Further investigation indicated that netrin-1 decreased the expression of Blood Vessel Epicardial Substance (BVES), which was down-regulated in HCC. Interestingly, LY294002, a special inhibitor to PI3K/AKT signaling which was determined as a downstream pathway of netrin-1, restored the reduction in BVES caused by netrin-1. In addition, BVES exhibited an opposite effect on HCC cell metastasis to that of netrin-1. Importantly, up-regulating BVES expression significantly attenuated netrin-1-enhanced migration and invasion, whereas silencing BVES expression rescued the metastatic phenotype in netrin-1 knockdown cells. Moreover, netrin-1 expression was negatively correlated with BVES in HCC tissues and cell lines with different metastatic potential. Taken together, these results reveal that netrin-1 promotes HCC cell metastasis by regulating BVES expression via AKT activation.
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Ntn1