Merck
  • Magnetic beads as an extraction medium for simultaneous quantification of acetaminophen and structurally related compounds in human serum.
Magnetic beads as an extraction medium for simultaneous quantification of acetaminophen and structurally related compounds in human serum.
Drug testing and analysis (2014-09-19)
Caroline Bylda, Vanya Velichkova, Jens Bolle, Roland Thiele, Uwe Kobold, Dietrich A Volmer
ABSTRACT
This paper describes a sample preparation method that complements a previously published liquid chromatography-tandem mass spectrometry (LC-MS/MS) assay for acetaminophen and eight structurally-related compounds in human serum (C. Bylda, R. Thiele, U. Kobold, D.A. Volmer. Drug Test. Anal. 2014, 6, 451). The analytes (acetaminophen [APAP] + metabolites acetaminophen-glucuronide [APG], -cysteine [APC], -mercapturate [APM] and -cysteine [APC], structurally similar analogues phenacetin and p-phenetidine, as well as tricyclic antidepressants imipramine and amitryptiline) were extracted from serum using magnetized hyper-crosslinked polystyrene particles. The sample preparation protocol was developed by means of a design of experiments (DoE) statistical approach. Using three representative compounds from the analyte panel with different polarities (high, medium, and low), two screening designs were used to identify factors that exhibited significant impact on recovery of the analytes. These parameters were then optimized to permit extraction of the complete target panel exhibiting a broad range of chemical polarities. Liquid chromatographic separations were achieved by gradient elution using a pentafluorphenyl column with subsequent detection by electrospray ionization-triple quadrupole mass spectrometry in multiple reaction monitoring (MRM) mode. The method was linear over the range 0.1-100 µg/mL for APAP, APG, p-phenetidine and phenacetin, 0.03-50 µg/mL for APS, and 0.01-10 µg/mL for APM, APC, imipramine and amitriptyline, with R(2)  > 0.99. The assay exhibited good precision with CVs ranging from 2 to 9% for all analytes; the accuracy was assessed by comparing two LC-MS/MS methods using a set of 68 patient samples.
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
4-Ethoxyaniline, 98%
Sigma-Aldrich
Adenosine 5′-phosphosulfate sodium salt, ≥85%
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Phenacetin, ≥98.0% (HPLC)
Sigma-Aldrich
Melamine, 99%
Sigma-Aldrich
Aphidicolin from Nigrospora sphaerica, ≥98% (HPLC), powder
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Supelco
Acetaminophen solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Methanol solution, (Methanol:Dimethyl sulfoxide 1:1 (v/v))
Sigma-Aldrich
Potassium sulfate, BioUltra, ≥99.0% (AT)
Sigma-Aldrich
Potassium sulfate, 99.99% trace metals basis
Sigma-Aldrich
Ammonium persulfate, BioUltra, for molecular biology, ≥98.0% (RT)
Sigma-Aldrich
Ammonium persulfate, for molecular biology, suitable for electrophoresis, ≥98%
Sigma-Aldrich
Ammonium persulfate, BioXtra, ≥98.0%
Sigma-Aldrich
Potassium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium sulfate, suitable for plant cell culture
Sigma-Aldrich
N-Acetyl-L-cysteine, BioReagent, suitable for cell culture
SAFC
Potassium sulfate
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
N-Acetyl-L-cysteine, Sigma Grade, ≥99% (TLC), powder
Sigma-Aldrich
Acetaminophen, meets USP testing specifications, 98.0-102.0%, powder
Sigma-Aldrich
Acetaminophen, BioXtra, ≥99.0%
Sigma-Aldrich
Acetaminophen, analytical standard