Merck
  • Effect of citrulline, urea, ethanol, and urease on the formation of ethyl carbamate in soybean paste model system.
Effect of citrulline, urea, ethanol, and urease on the formation of ethyl carbamate in soybean paste model system.
Food chemistry (2015-07-21)
Yong Gun Kim, Jihye Lyu, Mina K Kim, Kwang-Geun Lee
ABSTRACT
The aim of this study was to determine the effect of urease on the formation of ethyl carbamate (EC) in the presence of previously known precursors of EC (citrulline, urea, and ethanol) using a soybean paste model system. The levels of EC were quantitatively determined by gas chromatography-mass spectrometry (GC-MS) every five days for a 30-day period. After 30 days fermentation, the concentration of EC increased significantly by 135.2%, 242.2%, and 3757.1% when the precursors (citrulline, urea and ethanol) were added to the model system, respectively (p<0.05). Urease significantly decreased the level of EC by 38.4%, 18.8%, and 17.3% when citrulline, urea, and ethanol were added to the model system, respectively (p<0.05).
Sigma-Aldrich
Aluminum oxide, mesoporous, MSU-X (wormhole), average pore size 3.8 nm
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium sulfate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Water, for molecular biology, sterile filtered
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Water, BioPerformance Certified
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Aluminum oxide, nanopowder, 13 nm primary particle size (TEM), 99.8% trace metals basis
Sigma-Aldrich
Aluminum oxide, nanoparticles, <50 nm particle size (DLS), 20 wt. % in isopropanol
Sigma-Aldrich
Aluminum oxide, 99.997% trace metals basis
Sigma-Aldrich
Aluminum oxide, nanowires, diam. × L 2-6 nm × 200-400 nm
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Aluminum oxide, single crystal substrate, <0001>
Sigma-Aldrich
Aluminum oxide, nanoparticles, 30-60 nm particle size (TEM), 20 wt. % in H2O