Merck
Get up to 22% off for Pi Day until 3/26.Save Now
  • A novel four transmembrane spanning protein, CLP24. A hypoxically regulated cell junction protein.
A novel four transmembrane spanning protein, CLP24. A hypoxically regulated cell junction protein.
European journal of biochemistry (2004-06-23)
Jonathan Kearsey, Silvere Petit, Catherine De Oliveira, Fabien Schweighoffer
ABSTRACT
A novel hypoxically regulated intercellular junction protein (claudin-like protein of 24 kDa, CLP24) has been identified that shows homology to the myelin protein 22/epithelial membrane protein 1/claudin family of cell junction proteins, which are involved in the modulation of paracellular permeability. The CLP24 protein contains four predicted transmembrane domains and a C-terminal protein-protein interaction domain. These domains are characteristic of the four transmembrane spanning (tetraspan) family of proteins, which includes myelin protein 22, and are involved in cell adhesion at tight, gap and adherens junctions. Expression profiling analyses show that CLP24 is highly expressed in lung, heart, kidney and placental tissues. Cellular studies confirm that the CLP24 protein localizes to cell-cell junctions and co-localizes with the beta-catenin adherens junction-associated protein but not with tight junctions. Over-expression of CLP24 results in decreased adhesion between cells, and functional paracellular flux studies confirm that over-expression of the CLP24 protein modulates the junctional barrier function. These data therefore suggest that CLP24 is a novel, hypoxically regulated tetraspan adherens junction protein that modulates cell adhesion, paracellular permeability and angiogenesis.