Merck
  • Protein phosphatase 2A acts as a mitogen-activated protein kinase kinase kinase 3 (MEKK3) phosphatase to inhibit lysophosphatidic acid-induced IkappaB kinase beta/nuclear factor-kappaB activation.
Protein phosphatase 2A acts as a mitogen-activated protein kinase kinase kinase 3 (MEKK3) phosphatase to inhibit lysophosphatidic acid-induced IkappaB kinase beta/nuclear factor-kappaB activation.
The Journal of biological chemistry (2010-05-08)
Wenjing Sun, Hao Wang, Xiumei Zhao, Yang Yu, Yihui Fan, Hao Wang, Xishan Wang, Xiongbin Lu, Guiyin Zhang, Songbin Fu, Jianhua Yang
ABSTRACT
MEKK3 is a central intermediate signaling component in lysophosphatidic acid (LPA)-induced activation of the nuclear factor-kappaB (NF-kappaB). However, the precise mechanism for the termination of MEKK3 kinase activity is not fully understood. Using a functional genomic approach, we have identified a protein serine/threonine phosphatase, protein phosphatase 2A (PP2A), as a MEKK3 phosphatase. Overexpression of PP2A catalytic subunit (PP2Ac) beta-isoform results in dephosphorylation of MEKK3 at Thr-516 and Ser-520 and termination of MEKK3-mediated NF-kappaB activation. PP2Ac associates with the phosphorylated form of MEKK3 and the interaction between PP2Ac and MEKK3 is induced by LPA in a transient fashion in the cells. Furthermore, knockdown of PP2Ac expression enhances LPA-induced MEKK3-mediated IkappaB kinase beta (IKKbeta) phosphorylation and NF-kappaB activation. These data suggest that PP2A plays an important role in the termination of LPA-mediated NF-kappaB activation through dephosphorylating and inactivating MEKK3.