Merck
  • Inhibitory motor neurons of the esophageal myenteric plexus are mechanosensitive.
Inhibitory motor neurons of the esophageal myenteric plexus are mechanosensitive.
American journal of physiology. Cell physiology (2014-12-30)
Hui Dong, Yanfen Jiang, Jane Dong, Ravinder K Mittal
ABSTRACT
Mechanosensitivity of enteric neurons has been reported in the small intestine and colon, but not in the esophagus. Our earlier in vivo studies show that mechanical stretch of the esophagus in the axial direction induces neurally mediated relaxation of the lower esophageal sphincter, possibly through mechanosensitive motor neurons. However, this novel notion that the motor neurons are mechanosensitive has not been examined in isolated esophageal myenteric motor neurons. The goal of our present study was to examine the mechanosensitivity of esophageal motor neurons in primary culture and elucidate the underlying molecular mechanisms. Immmunocytochemical analysis revealed that >95% cells were positive for the neuronal marker protein gene product 9.5 and that 66% of these cells costained with protein gene product 9.5 and neuronal nitric oxide (NO) synthase. Hypotonic solution induced an increase in the cytoplasm volume in all cells that was independent of extracellular Ca(2+). Hypotonic solution and mechanical stretch induced cytoplasmic free Ca(2+) signaling in ~65% of neurons in the presence, but not absence, of extracellular Ca(2+). Neurons grown on the elastic membrane responded to mechanical stretch by an increase in neuronal size and Ca(2+) signaling simultaneously. Hypotonic stretch-induced cytoplasmic free Ca(2+) signaling was not affected by extracellular Mg(2+), 5-nitro-2-(3-phenylpropylamino)benzoic acid, and nifedipine but was attenuated by 2-aminoethoxydiphenyl borate, Gd(3+), and Grammostola mechanotoxin 4, blockers of the stretch-activated ion channels. In ~57% of the neurons, hypotonic stretch also induced Ca(2+)-dependent cytoplasmic NO production, which was abolished by Grammostola mechanotoxin 4. These results prove that the esophageal inhibitory motor neurons possess a mechanosensitive property and also provide novel insights into the stretch-activated ion channel-Ca(2+)-NO signaling pathway in these neurons.