Merck
  • Cosmomycin C inhibits signal transducer and activator of transcription 3 (STAT3) pathways in MDA-MB-468 breast cancer cell.
Cosmomycin C inhibits signal transducer and activator of transcription 3 (STAT3) pathways in MDA-MB-468 breast cancer cell.
Bioorganic & medicinal chemistry (2011-11-11)
Jihoon Kim, Yu-Jin Lee, Dae-Seop Shin, Sun-Hee Jeon, Kwang-Hee Son, Dong Cho Han, Seung-Nam Jung, Tae-Kwang Oh, Byoung-Mog Kwon
ABSTRACT
The signal transducer and activator of transcription 3 (STAT3) is constitutively activated in cancer cells. Therefore, blocking the aberrant activity of STAT3 in tumor cells is a validated therapeutic strategy. To discover novel inhibitors of STAT3 activity, we screened against microbial natural products using a dual-luciferase assay. Using the microbial metabolome library, we identified cosmomycin C (CosC), which was isolated from the mycelium extract of Streptomyces sp. KCTC19769, as a STAT3 pathway inhibitor. CosC inhibited STAT3 (Tyr705) phosphorylation and subsequent nuclear translocation in MDA-MB-468 breast cancer cells. CosC-mediated inhibition of STAT3 signaling pathway was confirmed by suppressed expression of STAT3 downstream target proteins including cyclin D1, Bcl-xL, survivin, Mcl-1, and VEGF in CosC-treated MDA-MB-468 cells. Flow cytometry showed that CosC caused accumulation in the G(0)-G(1) phase of the cell cycle and induced apoptosis via PARP cleavage and caspase-3 activation. Based on these findings, CosC may be a potential candidate for modulation of STAT3 pathway.
Product Description
Roche
X-tremeGENE HP DNA Transfection Reagent, High-performance polymer reagent for transfecting many cell lines