MilliporeSigma
  • Reversion-induced LIM interaction with Src reveals a novel Src inactivation cycle.
Reversion-induced LIM interaction with Src reveals a novel Src inactivation cycle.
The Journal of cell biology (2009-03-25)
Yongjun Zhang, Yizeng Tu, Jianping Zhao, Ka Chen, Chuanyue Wu
ABSTRACT
Aberrant Src activation plays prominent roles in cancer progression. However, how Src is activated in cancer cells is largely unknown. Genetic Src-activating mutations are rare and, therefore, are insufficient to account for Src activation commonly found in human cancers. In this study, we show that reversion-induced LIM (RIL), which is frequently lost in colon and other cancers as a result of epigenetic silencing, suppresses Src activation. Mechanistically, RIL suppresses Src activation through interacting with Src and PTPL1, allowing PTPL1-dependent dephosphorylation of Src at the activation loop. Importantly, the binding of RIL to Src is drastically reduced upon Src inactivation. Our results reveal a novel Src inactivation cycle in which RIL preferentially recognizes active Src and facilitates PTPL1-mediated inactivation of Src. Inactivation of Src, in turn, promotes dissociation of RIL from Src, allowing the initiation of a new Src inactivation cycle. Epigenetic silencing of RIL breaks this Src inactivation cycle and thereby contributes to aberrant Src activation in human cancers.