- Association of SWAP-70 with the B cell antigen receptor complex.
Association of SWAP-70 with the B cell antigen receptor complex.
Proceedings of the National Academy of Sciences of the United States of America (2000-02-19)
L Masat, J Caldwell, R Armstrong, H Khoshnevisan, R Jessberger, B Herndier, M Wabl, D Ferrick
PMID10681448
ABSTRACT
SWAP-70 is a component of an enzyme complex that recombines Ig switch regions in vitro. We report here the cloning of the human cDNA and its B lymphocyte-specific expression. Although its sequence contains three nuclear localization signals, in small resting B cells, SWAP-70 is mainly found in the cytoplasm. On stimulation, SWAP-70 translocates to the nucleus. In activated, class-switching B cell cultures, it is associated with membrane IgG, but not IgM. The membrane Ig association requires a functional pleckstrin homology domain and is controlled by the C terminus. We suggest that SWAP-70 is involved not only in nuclear events but also in signaling in B cell activation.