- Silver-catalyzed radical phosphonofluorination of unactivated alkenes.
Silver-catalyzed radical phosphonofluorination of unactivated alkenes.
Journal of the American Chemical Society (2013-09-13)
Chengwei Zhang, Zhaodong Li, Lin Zhu, Limei Yu, Zhentao Wang, Chaozhong Li
PMID24025164
ABSTRACT
We report herein a mild and catalytic phosphonofluorination of unactivated alkenes. With catalysis by AgNO3, the condensation of various unactivated alkenes with diethyl phosphite and Selectfluor reagent in CH2Cl2/H2O/HOAc at 40 °C led to the efficient synthesis of β-fluorinated alkylphosphonates with good stereoselectivity and wide functional group compatibility. A mechanism involving silver-catalyzed oxidative generation of phosphonyl radicals and silver-assisted fluorine atom transfer is proposed.