- Asymmetric synthesis of conformationally constrained fingolimod analogues--discovery of an orally active sphingosine 1-phosphate receptor type-1 agonist and receptor type-3 antagonist.
Asymmetric synthesis of conformationally constrained fingolimod analogues--discovery of an orally active sphingosine 1-phosphate receptor type-1 agonist and receptor type-3 antagonist.
Journal of medicinal chemistry (2007-11-13)
Ran Zhu, Ashley H Snyder, Yugesh Kharel, Lisa Schaffter, Qin Sun, Perry C Kennedy, Kevin R Lynch, Timothy L Macdonald
PMID17994678
ABSTRACT
Compound 1 (FTY720, Fingolimod) represents a new generation of immunosuppressant that modulates lymphocyte trafficking by interacting with the S1P(1) receptor. Compound 1 also provides a template molecule for studying the molecular biology of S1P receptors and related enzymes (kinases and phosphatases). In this study, two conformationally constrained analogues of 1 ( 3a and 3c) were asymmetrically synthesized in high optical purity. In vitro assessment documented that both analogues are Sphk2 substrates, their phosphorylated species are potent S1P(1) receptor agonists, and 3a-P is a potent S1P 3 antagonist. After oral administration in mice, both compounds evoked lymphopenia, but their duration of action differed markedly.