Merck
2-Methyl-5-nitro-benzonitrile.
Acta crystallographica. Section E, Structure reports online (2008-01-01)
Wen-Xian Liang, Guo-Xi Wang
ABSTRACT
In the title compound, C(8)H(6)N(2)O(2), the nitro group is rotated by 10.2 (2)° out of the plane of the benzene ring. The crystal structure is stabilized by van der Waals inter-actions.