Merck
  • 2-Chloro-2,2-difluoroacetophenone: a non-ODS-based difluorocarbene precursor and its use in the difluoromethylation of phenol derivatives.
2-Chloro-2,2-difluoroacetophenone: a non-ODS-based difluorocarbene precursor and its use in the difluoromethylation of phenol derivatives.
The Journal of organic chemistry (2006-12-16)
Laijun Zhang, Ji Zheng, Jinbo Hu
ABSTRACT
A novel and non-ODS-based (ODS = ozone-depleting substance) preparation of 2-chloro-2,2-difluoroacetophenone (1) was achieved in high yield by using 2,2,2-trifluoroacetophenone as the starting material. Compound 1 was found to act as a good difluorocarbene reagent, which readily reacts with a variety of structurally diverse phenol derivatives 4 in the presence of potassium hydroxide or potassium carbonate to produce aryl difluoromethyl ethers 5 in good yields. This new and easy-to-handle synthetic methodology offers an environmentally friendly alternative to other Freon- or Halon-based difluoromethylating approaches.
Sigma-Aldrich
2-Chloro-2,2-difluoroacetophenone, 95%